Product Details

SNP ID
rs10512330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104535936 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTCCCCAATCAGGTCTTGAGAC[G/T]TCGGTTTCGCATACATAAAGAAGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR13C3 PubMed Links

Gene Details

Gene
OR13C3
Gene Name
olfactory receptor family 13 subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001961.1 878 Missense Mutation AAG,ACG K293T NP_001001961.1
Gene
OR13C4
Gene Name
olfactory receptor family 13 subfamily C member 4
There are no transcripts associated with this gene.

View Full Product Details