Product Details

SNP ID
rs6689016
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:213067726 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTCCAGGAGGAAAGGGACTTGCT[C/T]ACCATTGTAATCCAGCACATAGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPS6KC1 PubMed Links

Gene Details

Gene
RPS6KC1
Gene Name
ribosomal protein S6 kinase C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136138.2 Intron NP_001129610.1
NM_001287218.1 Intron NP_001274147.1
NM_001287219.1 Intron NP_001274148.1
NM_001287220.1 Intron NP_001274149.1
NM_001287221.1 Intron NP_001274150.1
NM_012424.4 Intron NP_036556.2
XM_005273095.1 Intron XP_005273152.1
XM_005273096.1 Intron XP_005273153.1
XM_006711272.1 Intron XP_006711335.1
XM_017001024.1 Intron XP_016856513.1
XM_017001025.1 Intron XP_016856514.1
XM_017001026.1 Intron XP_016856515.1
XM_017001027.1 Intron XP_016856516.1
XM_017001028.1 Intron XP_016856517.1
XM_017001029.1 Intron XP_016856518.1
XM_017001030.1 Intron XP_016856519.1
XM_017001031.1 Intron XP_016856520.1
XM_017001032.1 Intron XP_016856521.1
XM_017001033.1 Intron XP_016856522.1
XM_017001034.1 Intron XP_016856523.1

View Full Product Details