Product Details

SNP ID
rs8182253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64040529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGAAAAGAATTGCCAAAGATAA[C/T]TGAATCGCATGCACACAAAACCTTA
Phenotype
MIM: 604033
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERN1 PubMed Links

Gene Details

Gene
ERN1
Gene Name
endoplasmic reticulum to nucleus signaling 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001433.3 7860 Intron NP_001424.3
XM_017024347.1 7860 UTR 3 XP_016879836.1
XM_017024348.1 7860 Intron XP_016879837.1

View Full Product Details