Product Details

SNP ID
rs6568951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116610861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTATCAGAGGATTCCGAAAGGTA[A/G]AGAAAAGAAGGAAGACTGGCCAGAG
Phenotype
MIM: 612647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RSPH4A PubMed Links

Gene Details

Gene
RSPH4A
Gene Name
radial spoke head 4 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010892.2 Intron NP_001010892.1
NM_001161664.1 Intron NP_001155136.1
XM_017010826.1 Intron XP_016866315.1

View Full Product Details