Product Details

SNP ID
rs10043606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150219528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGCTATTTATTGCACTGAAAAC[A/C]CCAAGAATAAAACAAACGCCCCTTC
Phenotype
MIM: 114078 MIM: 606205
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAMK2A PubMed Links

Gene Details

Gene
CAMK2A
Gene Name
calcium/calmodulin dependent protein kinase II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015981.3 4834 UTR 3 NP_057065.2
NM_171825.2 4834 UTR 3 NP_741960.1
XM_017009898.1 4834 Intron XP_016865387.1
Gene
SLC6A7
Gene Name
solute carrier family 6 member 7
There are no transcripts associated with this gene.

View Full Product Details