Product Details

SNP ID
rs6017704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45857946 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCGTCTCCGTCCGCTTCCACAAC[C/T]TCAACCATGGCACCTCCATCCGCGA
Phenotype
MIM: 608123
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOT8 PubMed Links

Gene Details

Gene
ACOT8
Gene Name
acyl-CoA thioesterase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005469.3 366 Intron NP_005460.2
XM_005260239.3 366 Intron XP_005260296.1
XM_011528480.2 366 Intron XP_011526782.1
Gene
ZSWIM3
Gene Name
zinc finger SWIM-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080752.3 366 Missense Mutation CTC,TTC L41F NP_542790.2

View Full Product Details