Product Details

SNP ID
rs10431138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62271911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGTAGATCATGGGTATGAAACC[A/C]CCATGGAGGAAGAATTGCCTCAAAC
Phenotype
MIM: 605562
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCGB2A2 PubMed Links

Gene Details

Gene
SCGB2A2
Gene Name
secretoglobin family 2A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002411.3 Intron NP_002402.1
XM_005274005.3 Intron XP_005274062.3

View Full Product Details