Product Details

SNP ID
rs6104473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46120388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAACTAGTAAGTGTTACAAAGTCA[A/G]GACTGGAGTCTAAAGCTGTCTGACT
Phenotype
MIM: 109535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD40 PubMed Links

Gene Details

Gene
CD40
Gene Name
CD40 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001250.5 Intron NP_001241.1
NM_001302753.1 Intron NP_001289682.1
NM_001322421.1 Intron NP_001309350.1
NM_001322422.1 Intron NP_001309351.1
NM_152854.3 Intron NP_690593.1

View Full Product Details