Product Details

SNP ID
rs10157137
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:35435621 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTAGGGTGGCAGAGTAGGTGCAC[A/G]CTTGGACAGGTCTCTCCTGCTGGGA
Phenotype
MIM: 613535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0319L PubMed Links

Gene Details

Gene
KIAA0319L
Gene Name
KIAA0319 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024874.4 Intron NP_079150.3
XM_006710907.1 Intron XP_006710970.1
XM_006710909.1 Intron XP_006710972.1
XM_006710910.1 Intron XP_006710973.1
XM_011542179.2 Intron XP_011540481.1
XM_011542180.1 Intron XP_011540482.1
XM_017002366.1 Intron XP_016857855.1
XM_017002367.1 Intron XP_016857856.1
XM_017002368.1 Intron XP_016857857.1
XM_017002369.1 Intron XP_016857858.1
XM_017002370.1 Intron XP_016857859.1
XM_017002371.1 Intron XP_016857860.1
XM_017002372.1 Intron XP_016857861.1
XM_017002373.1 Intron XP_016857862.1
XM_017002374.1 Intron XP_016857863.1
XM_017002375.1 Intron XP_016857864.1
XM_017002376.1 Intron XP_016857865.1
XM_017002377.1 Intron XP_016857866.1
XM_017002378.1 Intron XP_016857867.1

View Full Product Details