Product Details

SNP ID
rs9901197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1429260 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTCCCAGCTACTTGGGAGACTGA[A/G]GGGAGAGGAACCCTTGAGTCCAGGG
Phenotype
MIM: 164762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRK PubMed Links

Gene Details

Gene
CRK
Gene Name
CRK proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005206.4 Intron NP_005197.3
NM_016823.3 Intron NP_058431.2

View Full Product Details