Product Details

SNP ID
rs10198300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74199537 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTGATGGTTGCACAACATATGAA[C/T]GTATTTAATACCACTGAACTGCACT
Phenotype
MIM: 604887
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTHFD2 PubMed Links

Gene Details

Gene
MTHFD2
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006636.3 Intron NP_006627.2
XM_006711924.2 Intron XP_006711987.1

View Full Product Details