Product Details

SNP ID
rs6102213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36778428 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGTCTTGGGTGGGGCTGCTAAT[C/T]TGCACTTCTATCAAGTTCCCAGGTG
Phenotype
MIM: 609175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSN1 PubMed Links

Gene Details

Gene
DSN1
Gene Name
DSN1 homolog, MIS12 kinetochore complex component
There are no transcripts associated with this gene.

Gene
SOGA1
Gene Name
suppressor of glucose, autophagy associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080627.3 13383 UTR 3 NP_542194.2
NM_199181.2 13383 Intron NP_954650.2

View Full Product Details