Product Details
- SNP ID
-
rs9624364
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:23969285 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTCACCTCTGAGAGGGGCATGTTG[A/G]CTTATGGCACAGACATTAGTGGTGG
- Phenotype
-
MIM: 602750
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DDT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76469591] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDT
- Gene Name
- D-dopachrome tautomerase
There are no transcripts associated with this gene.
- Gene
- DDTL
- Gene Name
- D-dopachrome tautomerase-like
- Gene
- GSTT2B
- Gene Name
- glutathione S-transferase theta 2B (gene/pseudogene)
There are no transcripts associated with this gene.
- Gene
- LOC107985559
- Gene Name
- uncharacterized LOC107985559
There are no transcripts associated with this gene.
View Full Product Details