Product Details

SNP ID
rs10408452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40752450 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCCGTATAGAATTCTGCTTAGGA[A/G]CCCAATTTTTGGCCGGGCGTGGTGG
Phenotype
MIM: 182285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
There are no transcripts associated with this gene.

Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004596.4 Intron NP_004587.1

View Full Product Details