Product Details

SNP ID
rs6011981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46541718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCCGCAGGCGGCGGGCGTAGGC[C/T]TCCAGGAGCACCGTGGAGCCCGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OCSTAMP PubMed Links

Gene Details

Gene
OCSTAMP
Gene Name
osteoclast stimulatory transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080721.2 1271 Silent Mutation GAA,GAG E419E NP_542452.1

View Full Product Details