Product Details

SNP ID
rs9436297
Assay Type
Functionally Tested
NCBI dbSNP Submissions
54
Location
Chr.1:65423171 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGGTCAAGGCAGAGAAGTCCAGAC[C/T]GGAGGTGGAAGTACAGGCCTAGTTC
Phenotype
MIM: 608375 MIM: 601007 MIM: 613461
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DNAJC6 PubMed Links

Gene Details

Gene
DNAJC6
Gene Name
DnaJ heat shock protein family (Hsp40) member C6
There are no transcripts associated with this gene.

Gene
LEPR
Gene Name
leptin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003679.3 Intron NP_001003679.1
NM_001003680.3 Intron NP_001003680.1
NM_001198687.1 Intron NP_001185616.1
NM_001198688.1 Intron NP_001185617.1
NM_001198689.1 Intron NP_001185618.1
NM_002303.5 Intron NP_002294.2
Gene
LEPROT
Gene Name
leptin receptor overlapping transcript
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198681.1 Intron NP_001185610.1
NM_001198683.1 Intron NP_001185612.1
NM_017526.4 Intron NP_059996.1

View Full Product Details