Product Details

SNP ID
rs10095681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:24304148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGCTGTGAAGTGATTTCCTCA[C/T]GACTGTAAAAAATAAATTTGAGTTT
Phenotype
MIM: 606188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM28 PubMed Links

Gene Details

Gene
ADAM28
Gene Name
ADAM metallopeptidase domain 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304351.1 Intron NP_001291280.1
NM_014265.5 Intron NP_055080.2
NM_021777.4 Intron NP_068547.2
XM_005273380.3 Intron XP_005273437.1
XM_005273382.1 Intron XP_005273439.1
XM_006716273.3 Intron XP_006716336.1
XM_006716274.1 Intron XP_006716337.1
XM_011544367.2 Intron XP_011542669.1
XM_011544368.2 Intron XP_011542670.1
XM_011544369.2 Intron XP_011542671.1
XM_011544370.2 Intron XP_011542672.1
XM_011544371.2 Intron XP_011542673.1
XM_017012974.1 Intron XP_016868463.1
XM_017012975.1 Intron XP_016868464.1
XM_017012976.1 Intron XP_016868465.1
Gene
LOC101929294
Gene Name
uncharacterized LOC101929294
There are no transcripts associated with this gene.

View Full Product Details