Product Details

SNP ID
rs9304684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49388265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAGCTCACGCGCACCTCCCCCA[A/G]CTCAACTGCCGTTCGTCGTTCGTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC155 PubMed Links

Gene Details

Gene
CCDC155
Gene Name
coiled-coil domain containing 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144688.4 48 UTR 5 NP_653289.3
XM_005258549.3 48 UTR 5 XP_005258606.1
XM_011526488.2 48 UTR 5 XP_011524790.1
XM_011526489.2 48 Intron XP_011524791.1
XM_011526490.2 48 UTR 5 XP_011524792.1
XM_011526494.2 48 Intron XP_011524796.1
XM_011526495.2 48 Intron XP_011524797.1
XM_011526496.2 48 UTR 5 XP_011524798.1
XM_011526497.2 48 Intron XP_011524799.1
XM_017026333.1 48 Intron XP_016881822.1
Gene
LOC101928295
Gene Name
uncharacterized LOC101928295
There are no transcripts associated with this gene.

View Full Product Details