Product Details

SNP ID
rs6631192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30844042 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACAAAAAAGCTGGTTTTCATCAA[A/G]TCATGCACTGATTTGCATCTATTCT
Phenotype
MIM: 300480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAB3 PubMed Links

Gene Details

Gene
TAB3
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152787.4 Intron NP_690000.3
XM_005274483.3 Intron XP_005274540.2
XM_005274484.2 Intron XP_005274541.2
XM_011545488.2 Intron XP_011543790.1
XM_017029399.1 Intron XP_016884888.1
XM_017029400.1 Intron XP_016884889.1
XM_017029401.1 Intron XP_016884890.1
XM_017029402.1 Intron XP_016884891.1

View Full Product Details