Product Details

SNP ID
rs9383302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:17622509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTAACATCTTTACTATCCAGGT[C/T]TACCTTGTTATAGAACTGAGAACCT
Phenotype
MIM: 603948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUP153 PubMed Links

Gene Details

Gene
NUP153
Gene Name
nucleoporin 153
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278209.1 Intron NP_001265138.1
NM_001278210.1 Intron NP_001265139.1
NM_005124.3 Intron NP_005115.2
XM_005249507.2 Intron XP_005249564.1
XM_006715290.2 Intron XP_006715353.1
XM_006715291.3 Intron XP_006715354.1
XM_011515028.2 Intron XP_011513330.1
XM_017011594.1 Intron XP_016867083.1

View Full Product Details