Product Details

SNP ID
rs7324931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72716326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGAGGGGTTAGTGACTGGGGAG[A/G]GGGCAGAAGGGGGATTATCTGGTGC
Phenotype
MIM: 613448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MZT1 PubMed Links

Gene Details

Gene
MZT1
Gene Name
mitotic spindle organizing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001071775.2 Intron NP_001065243.1

View Full Product Details