Product Details
- SNP ID
-
rs5925970
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:23987107 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATACATGTTTCAGAAATATAAGGA[A/G]TTCAAATATTACTGAATCAGTATGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL15
PubMed Links
Gene Details
- Gene
- KLHL15
- Gene Name
- kelch like family member 15
View Full Product Details