Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001030059.2 | 11167 | Intron | NP_001025230.1 | ||
NM_001318166.1 | 11167 | Intron | NP_001305095.1 | ||
NM_001318167.1 | 11167 | Intron | NP_001305096.1 | ||
NM_001318168.1 | 11167 | Intron | NP_001305097.1 | ||
NM_001318169.1 | 11167 | Intron | NP_001305098.1 | ||
XM_005269592.2 | 11167 | UTR 3 | XP_005269649.1 | ||
XM_006717685.3 | 11167 | UTR 3 | XP_006717748.1 | ||
XM_006717686.3 | 11167 | Intron | XP_006717749.1 | ||
XM_011539444.2 | 11167 | UTR 3 | XP_011537746.1 | ||
XM_011539445.2 | 11167 | Intron | XP_011537747.1 | ||
XM_017015820.1 | 11167 | Intron | XP_016871309.1 | ||
XM_017015821.1 | 11167 | UTR 3 | XP_016871310.1 | ||
XM_017015822.1 | 11167 | UTR 3 | XP_016871311.1 | ||
XM_017015823.1 | 11167 | Intron | XP_016871312.1 | ||
XM_017015824.1 | 11167 | Intron | XP_016871313.1 |