Product Details
- SNP ID
-
hCV30279471
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:134572924 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCGAGTTTGTATTGTGTTTTACA[C/G]GGGAAAATATATTAAGCCTCACCTT
- Phenotype
-
MIM: 616336
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AKR1B15
PubMed Links
Gene Details
- Gene
- AKR1B15
- Gene Name
- aldo-keto reductase family 1 member B15
There are no transcripts associated with this gene.
View Full Product Details