Product Details
- SNP ID
-
rs6694765
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:157679036 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGGCAATATATTCCAACTTACAAC[C/G]TACGCACACTGCATTCCAAACCAAT
- Phenotype
-
MIM: 606510
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FCRL3
PubMed Links
Gene Details
- Gene
- FCRL3
- Gene Name
- Fc receptor like 3
View Full Product Details