Product Details

SNP ID
rs6970288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76220935 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAGGGTTGGGTAACAAACAGCC[A/G]TGCCAGCACCAGCGCCTCTCCAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRRM3 PubMed Links

Gene Details

Gene
SRRM3
Gene Name
serine/arginine repetitive matrix 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110199.2 Intron NP_001103669.1
NM_001291831.1 Intron NP_001278760.1

View Full Product Details