Product Details

SNP ID
rs4311633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:76982067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGAACTGGCAGTGTATTTCCA[C/T]CATACTAGCAAGATGTTTATTTAAA
Phenotype
MIM: 170993
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PEX2 PubMed Links

Gene Details

Gene
PEX2
Gene Name
peroxisomal biogenesis factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000318.2 2570 UTR 3 NP_000309.1
NM_001079867.1 2570 UTR 3 NP_001073336.1
NM_001172086.1 2570 UTR 3 NP_001165557.1
NM_001172087.1 2570 UTR 3 NP_001165558.1

View Full Product Details