Product Details

SNP ID
rs10419021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8364750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGCGCAACTGTGGCTCCCTGGG[C/T]TTCCCTGCGTCAAGGGATGGGCTCC
Phenotype
MIM: 605910
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANGPTL4 PubMed Links

Gene Details

Gene
ANGPTL4
Gene Name
angiopoietin like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039667.2 Intron NP_001034756.1
NM_139314.2 Intron NP_647475.1
XM_005272484.3 Intron XP_005272541.1
XM_005272485.3 Intron XP_005272542.1
Gene
RAB11B-AS1
Gene Name
RAB11B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details