Product Details

SNP ID
rs9349153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:12259203 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATGCATTAAATCATGAAATAGAA[A/G]TGAAGTATAATTCCTTAAGGAGAGG
Phenotype
MIM: 131240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDN1 PubMed Links

Gene Details

Gene
EDN1
Gene Name
endothelin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168319.1 Intron NP_001161791.1
NM_001955.4 Intron NP_001946.3
XM_011514330.2 Intron XP_011512632.1
XM_011514331.2 Intron XP_011512633.1
XM_011514332.2 Intron XP_011512634.1
XM_017010331.1 Intron XP_016865820.1

View Full Product Details