Product Details

SNP ID
rs10208498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:214933778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTACCATGGAAGAAGGAAGGAAA[A/G]TGTTAACAGCTGTTTGTTATGTATG
Phenotype
MIM: 607800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA12 PubMed Links

Gene Details

Gene
ABCA12
Gene Name
ATP binding cassette subfamily A member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015657.3 Intron NP_056472.2
NM_173076.2 Intron NP_775099.2
XM_011510951.2 Intron XP_011509253.1
Gene
LOC101928103
Gene Name
uncharacterized LOC101928103
There are no transcripts associated with this gene.

View Full Product Details