Product Details
- SNP ID
-
rs7546625
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
39
- Location
-
Chr.1:84086923 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGCCTGTTGGTCCTCTGAGCAGGG[A/G]GCTTAACTGTGAACAAATCAGACAA
- Phenotype
-
MIM: 176892
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRKACB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80179969] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRKACB
- Gene Name
- protein kinase cAMP-activated catalytic subunit beta
View Full Product Details