Product Details

SNP ID
rs10275990
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:71782150 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAGAGGCTGGATATTTGTTCCCC[C/T]CAAATTTCATGTTGATCTCCAGTGC
Phenotype
MIM: 607176
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CALN1 PubMed Links

Gene Details

Gene
CALN1
Gene Name
calneuron 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017440.2 6601 UTR 3 NP_001017440.1
NM_031468.3 6601 UTR 3 NP_113656.2
XM_011516594.2 6601 UTR 3 XP_011514896.1
XM_011516595.1 6601 UTR 3 XP_011514897.1
XM_011516596.2 6601 UTR 3 XP_011514898.1
XM_011516597.2 6601 UTR 3 XP_011514899.1
XM_011516599.1 6601 UTR 3 XP_011514901.1
XM_017012675.1 6601 UTR 3 XP_016868164.1
XM_017012676.1 6601 UTR 3 XP_016868165.1
XM_017012677.1 6601 UTR 3 XP_016868166.1
XM_017012678.1 6601 UTR 3 XP_016868167.1
XM_017012679.1 6601 UTR 3 XP_016868168.1
XM_017012680.1 6601 UTR 3 XP_016868169.1
XM_017012681.1 6601 UTR 3 XP_016868170.1
XM_017012682.1 6601 UTR 3 XP_016868171.1
XM_017012683.1 6601 UTR 3 XP_016868172.1

View Full Product Details