Product Details

SNP ID
rs9503233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2623696 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATGCTGTAGCCCCCTTAGAGCGG[A/G]CCTGGCTCCTGGCTGGCAGAGTGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01600 PubMed Links

Gene Details

Gene
LINC01600
Gene Name
long intergenic non-protein coding RNA 1600
There are no transcripts associated with this gene.

View Full Product Details