Product Details

SNP ID
rs4766262
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4621920 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACAACAACAAAACACCAATCAAA[A/G]GACCAATGGCAAAGGAAACATTTGT
Phenotype
MIM: 604689 MIM: 609181
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AKAP3 PubMed Links

Gene Details

Gene
AKAP3
Gene Name
A-kinase anchoring protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278309.1 Intron NP_001265238.1
NM_006422.3 Intron NP_006413.3
XM_005253664.2 Intron XP_005253721.1
XM_011520908.1 Intron XP_011519210.1
XM_011520909.1 Intron XP_011519211.1
XM_011520910.2 Intron XP_011519212.1
Gene
DYRK4
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 4
There are no transcripts associated with this gene.

View Full Product Details