Product Details

SNP ID
rs6028208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38927432 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGCTAATTACTTTCATGCATCTT[A/T]TGCTGTAAAAAGCAGATAAACACGA
Phenotype
MIM: 613275
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM83D PubMed Links

Gene Details

Gene
FAM83D
Gene Name
family with sequence similarity 83 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030919.2 Intron NP_112181.3
XM_017028088.1 Intron XP_016883577.1
Gene
PPP1R16B
Gene Name
protein phosphatase 1 regulatory subunit 16B
There are no transcripts associated with this gene.

View Full Product Details