Product Details

SNP ID
rs9947834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:52345488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGAATCATAGCTTGGAATCTCAC[C/T]AACTCATGATTAATAGACACTCAAT
Phenotype
MIM: 120470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCC PubMed Links

Gene Details

Gene
DCC
Gene Name
DCC netrin 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005215.3 Intron NP_005206.2
XM_011525843.1 Intron XP_011524145.1
XM_011525844.2 Intron XP_011524146.1
XM_017025568.1 Intron XP_016881057.1
XM_017025569.1 Intron XP_016881058.1
XM_017025570.1 Intron XP_016881059.1

View Full Product Details