Product Details

SNP ID
rs6071780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36656631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAACAGAACAAGATTGAATTCTA[A/G]GTTTTAATGTTTAACTGCAAGTTTG
Phenotype
MIM: 605273
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDRG3 PubMed Links

Gene Details

Gene
NDRG3
Gene Name
NDRG family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022477.3 Intron NP_071922.2
NM_032013.3 Intron NP_114402.1

View Full Product Details