Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_017698.2 | 5508 | Intron | NP_060168.2 | ||
NM_032227.3 | 5508 | Intron | NP_115603.2 | ||
XM_005262205.4 | 5508 | Intron | XP_005262262.1 | ||
XM_005262206.3 | 5508 | Intron | XP_005262263.1 | ||
XM_005262208.4 | 5508 | Intron | XP_005262265.1 | ||
XM_011531051.2 | 5508 | Intron | XP_011529353.1 | ||
XM_011531054.1 | 5508 | Intron | XP_011529356.1 | ||
XM_011531055.2 | 5508 | Intron | XP_011529357.1 | ||
XM_017029894.1 | 5508 | Intron | XP_016885383.1 | ||
XM_017029895.1 | 5508 | UTR 3 | XP_016885384.1 | ||
XM_017029896.1 | 5508 | Intron | XP_016885385.1 | ||
XM_017029897.1 | 5508 | Intron | XP_016885386.1 | ||
XM_017029898.1 | 5508 | Intron | XP_016885387.1 | ||
XM_017029899.1 | 5508 | Intron | XP_016885388.1 | ||
XM_017029900.1 | 5508 | Intron | XP_016885389.1 |