Product Details

SNP ID
rs6058147
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34963652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTCATATCTACGAAACCATTGC[C/T]TAATTCGAAGGTCACAAAGATTTAC
Phenotype
MIM: 601002 MIM: 609928
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GSS PubMed Links
Additional Information
For this assay, SNP(s) [rs80170004] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GSS
Gene Name
glutathione synthetase
There are no transcripts associated with this gene.

Gene
MYH7B
Gene Name
myosin heavy chain 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020884.4 Intron NP_065935.3
XM_006723840.3 Intron XP_006723903.1
XM_011528941.2 Intron XP_011527243.1
XM_011528947.2 Intron XP_011527249.1
XM_011528948.2 Intron XP_011527250.1
XM_017027986.1 Intron XP_016883475.1
XM_017027987.1 Intron XP_016883476.1

View Full Product Details