Product Details
- SNP ID
-
rs9472084
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:43585614 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAGTATCCTTTTCTGGCTGTGTGT[A/G]TGAGTATATTGCTCTTTCTTTTCTT
- Phenotype
-
MIM: 603968
MIM: 607845
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
POLH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201521491] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POLH
- Gene Name
- DNA polymerase eta
- Gene
- XPO5
- Gene Name
- exportin 5
There are no transcripts associated with this gene.
View Full Product Details