Product Details

SNP ID
rs5934788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:10169483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATGAGTGAAAAGTAACAAGGGA[A/G]ACAAGATGAATTCAGAAAAACAAAC
Phenotype
MIM: 302910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCN4 PubMed Links

Gene Details

Gene
CLCN4
Gene Name
chloride voltage-gated channel 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256944.1 Intron NP_001243873.1
NM_001830.3 Intron NP_001821.2

View Full Product Details