Product Details

SNP ID
rs10461962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36197907 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACCCACAATACAGTAGGAATAGG[C/T]ACAACTGGGGAAAAAGAAAAAAATA
Phenotype
MIM: 615787 MIM: 601436
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NADK2 PubMed Links

Gene Details

Gene
NADK2
Gene Name
NAD kinase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085411.2 Intron NP_001078880.1
NM_001287340.1 Intron NP_001274269.1
NM_001287341.1 Intron NP_001274270.1
NM_153013.4 Intron NP_694558.1
XM_005248241.4 Intron XP_005248298.1
Gene
SKP2
Gene Name
S-phase kinase-associated protein 2, E3 ubiquitin protein ligase
There are no transcripts associated with this gene.

View Full Product Details