Product Details
- SNP ID
-
rs9912981
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:39985383 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAACGTGATGAACGGATTGGGAAG[C/T]GGGCAGTATAAAGCTCCTAGAGCAG
- Phenotype
-
MIM: 611218
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GSDMA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78767867] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GSDMA
- Gene Name
- gasdermin A
There are no transcripts associated with this gene.
- Gene
- PSMD3
- Gene Name
- proteasome 26S subunit, non-ATPase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002809.3 |
|
Intron |
|
|
NP_002800.2 |
View Full Product Details