Product Details

SNP ID
rs10417732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2824047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTAGGACTTGGCCCAAACCCCA[C/T]TCTGAGAGAGGCCAGGCAGTGGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF554 PubMed Links

Gene Details

Gene
ZNF554
Gene Name
zinc finger protein 554
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102651.1 Intron NP_001096121.1
XM_006722635.3 Intron XP_006722698.1
XM_011527660.2 Intron XP_011525962.1
XM_011527662.2 Intron XP_011525964.1
XM_011527664.2 Intron XP_011525966.1
XM_011527667.2 Intron XP_011525969.1
XM_017026234.1 Intron XP_016881723.1
XM_017026235.1 Intron XP_016881724.1
XM_017026236.1 Intron XP_016881725.1

View Full Product Details