Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001323374.1 | 2471 | Intron | NP_001310303.1 | ||
NM_001323375.1 | 2471 | Intron | NP_001310304.1 | ||
NM_001323376.1 | 2471 | Intron | NP_001310305.1 | ||
NM_001323377.1 | 2471 | Intron | NP_001310306.1 | ||
NM_001744.5 | 2471 | Intron | NP_001735.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001308056.1 | 2471 | UTR 3 | NP_001294985.1 | ||
NM_001308057.1 | 2471 | Intron | NP_001294986.1 | ||
NM_001308058.1 | 2471 | Intron | NP_001294987.1 | ||
NM_001308059.1 | 2471 | UTR 3 | NP_001294988.1 | ||
NM_001308060.1 | 2471 | UTR 3 | NP_001294989.1 | ||
NM_001308061.1 | 2471 | UTR 3 | NP_001294990.1 | ||
NM_139164.2 | 2471 | UTR 3 | NP_631903.1 | ||
XM_017009041.1 | 2471 | UTR 3 | XP_016864530.1 | ||
XM_017009042.1 | 2471 | UTR 3 | XP_016864531.1 | ||
XM_017009043.1 | 2471 | UTR 3 | XP_016864532.1 | ||
XM_017009044.1 | 2471 | UTR 3 | XP_016864533.1 |