Product Details

SNP ID
rs6091784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:53944008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATCTTTGCCGTTGTCACTGAGC[C/T]TCAAAAGCAATTGTTTTCCCAAATC
Phenotype
MIM: 602968
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCAS1 PubMed Links

Gene Details

Gene
BCAS1
Gene Name
breast carcinoma amplified sequence 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316361.2 2773 UTR 3 NP_001303290.1
NM_001323347.1 2773 UTR 3 NP_001310276.1
NM_003657.3 2773 UTR 3 NP_003648.2

View Full Product Details