Product Details
- SNP ID
-
rs10008985
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70075246 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATCATGTGTCCGTTTAAAACTTAA[A/C]CCTGAGCAATTCAAAAAACACATGT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CSN1S2AP
PubMed Links
Gene Details
- Gene
- CSN1S2AP
- Gene Name
- casein alpha s2-like A, pseudogene
There are no transcripts associated with this gene.
View Full Product Details