Product Details

SNP ID
rs3828411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195726120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCACTCCCCACTAACAGCGCCA[C/T]AGAAAGAGAAGTGACAGCACCCGGG
Phenotype
MIM: 610360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MUC20 PubMed Links

Gene Details

Gene
MUC20
Gene Name
mucin 20, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282506.1 1643 Missense Mutation ACA,ATA T506I NP_001269435.1
NM_001291833.1 1643 Missense Mutation ACA,ATA T332I NP_001278762.1
NM_020790.1 1643 Missense Mutation CTA,TTA L354L NP_065841.1
NM_152673.3 1643 Missense Mutation ACA,ATA T335I NP_689886.3

View Full Product Details