Product Details

SNP ID
rs6702340
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:196975920 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGAGTGTGTGCAAGGGACTGACT[G/T]ACAGTGATGAGCCATGGAATCTAAT
Phenotype
MIM: 608593
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CFHR5 PubMed Links

Gene Details

Gene
CFHR5
Gene Name
complement factor H related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030787.3 Intron NP_110414.1
XM_011510020.2 Intron XP_011508322.1

View Full Product Details