Product Details

SNP ID
rs9401591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:122754906 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCCTGGTGTCCCTTTCAATTACA[A/G]TGTTCTCCACCTAAACTCACCAACC
Phenotype
MIM: 602965
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FABP7 PubMed Links

Gene Details

Gene
FABP7
Gene Name
fatty acid binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319039.1 Intron NP_001305968.1
NM_001319041.1 Intron NP_001305970.1
NM_001319042.1 Intron NP_001305971.1
NM_001446.4 Intron NP_001437.1

View Full Product Details